Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ether, Ultra Low Water, Contains No Preservative, BakerDRY™, J.T. Baker™
Shop All Avantor Products
Click to view available options
Quantity:
100 mL
1 L
Packaging:
Glass Bottle
Narrow Mouth Amber Glass Bottle, w/ Septum
Specifications
Specifications
Flash Point | −45.00°C |
Packaging | Narrow Mouth Amber Glass Bottle, w/ Septum |
Quantity | 1 L |
Specific Gravity | 1L = 0.71kg |
Formula Weight | 74.12 |
Grade | BakerDRY™ |
Chemical Name or Material | Ether, Ultra Low Water, Contains No Preservative |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction