Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ether, Ultra Low Water, Contains No Preservative, BakerDRY™, J.T. Baker™

Catalog No. p-5391130
Click to view available options
Quantity:
100 mL
1 L
Packaging:
Glass Bottle
Narrow Mouth Amber Glass Bottle, w/ Septum
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifically manufactured for use in organometallic and oligonucleotide synthesis

Chemical Identifiers

CAS 60-29-7
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00011646
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
Synonym diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
PubChem CID 3283
ChEBI CHEBI:35702
IUPAC Name ethoxyethane
SMILES CCOCC

Specifications

Flash Point −45.00°C
Packaging Glass Bottle
Quantity 100 mL
Specific Gravity 1L = 0.71kg
Formula Weight 74.12
Grade BakerDRY™
Chemical Name or Material Ether, Ultra Low Water, Contains No Preservative
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.