Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ether, Ultra Low Water, Contains No Preservative, BakerDRY™, J.T.Baker™

Click to view available options
Quantity:
100mL
1L
Packaging:
Glass Bottle
Narrow Mouth Amber Glass Bottle, w/ Septum

Chemical Identifiers

60-29-7
C4H10O
74.123
RTZKZFJDLAIYFH-UHFFFAOYSA-N
3283
CHEBI:35702

Specifically manufactured for use in organometallic and oligonucleotide synthesis

SMILES CCOCC
IUPAC Name ethoxyethane
Formula Weight 74.12
Packaging Glass Bottle
Specific Gravity 1L = 0.71kg
Quantity 100mL
Flash Point −45.00°C
Chemical Name or Material Ether, Ultra Low Water, Contains No Preservative
Grade BakerDRY™