Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-amino-1-cyclohexene-1-carboxylate, 98%, Thermo Scientific Chemicals

Catalog No. AC334260050
Click to view available options
Quantity:
1 g
5 g

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1128-00-3
Molecular Formula C9H15NO2
Molecular Weight (g/mol) 169.22
MDL Number MFCD01863243
InChI Key JBZVWABPSHNPIK-UHFFFAOYSA-N
Synonym ethyl 2-amino-1-cyclohexene-1-carboxylate, ethyl 2-aminocyclohex-1-enecarboxylate, 2-amino-cyclohex-1-enecarboxylic acid ethyl ester, ethyl 2-aminocyclohex-1-ene-1-carboxylate, 2-amino-1-cyclohexenecarboxylic acid ethyl ester, 1-cyclohexene-1-carboxylic acid, 2-amino-, ethyl ester, 2-aminocyclohex-1-enecarboxylic acid ethyl ester, 1-cyclohexene-1-carboxylicacid, 2-amino-, ethyl ester, 2-amino-cyclohex-1-enecarboxylicacidethylester, ethyl cis-2-aminocyclohexenecarboxylate
PubChem CID 312929
SMILES CCOC(=O)C1=C(N)CCCC1

Specifications

Melting Point 70°C to 74°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Linear Formula NHC6H8COOCHCH3
Quantity 5 g
IUPAC Name ethyl 2-aminocyclohex-1-ene-1-carboxylate
Formula Weight 169.22
Percent Purity ≥97.5%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Ethyl 2-amino-1-cyclohexene-1-carboxylate, 98%
Show More Show Less
Hazard Category LIGHT SENSITIVE

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.