Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-chloro-4,4,4-trifluoroacetoacetate, 94%

Catalog No. aab2227006
Click to view available options
Quantity:
5 g
25 g

CAS: 363-58-6 | C6H6ClF3O3 | 218.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 363-58-6
Molecular Formula C6H6ClF3O3
Molecular Weight (g/mol) 218.56
MDL Number MFCD00041540
InChI Key YVWUNJVPOCYLIM-VKHMYHEASA-N
Synonym ethyl 2-chloro-4,4,4-trifluoroacetoacetate, ethyl 2-chloro-3-keto-4,4,4-trifluorobutyrate, ethyl 2-chloro-4,4,4-trifluoro-3-oxobutyrate, 2-chloro-4,4,4-trifluoro-3-oxo-butyric acid ethyl ester, 2-chloro-4,4,4-trifluoroacetoacetic acid ethyl ester, butanoic acid, 2-chloro-4,4,4-trifluoro-3-oxo-, ethyl ester, zlchem 1230, acmc-1cs5z, ethyl chlorotrifluoroacetoacetate, ethyl 2-chloro-3-keto-trifluorobutyrate
PubChem CID 2737165
IUPAC Name ethyl 2-chloro-4,4,4-trifluoro-3-oxobutanoate
SMILES CCOC(=O)[C@@H](Cl)C(=O)C(F)(F)F

Specifications

Density 1.39
Boiling Point 67°C to 71°C (35 mmHg)
Flash Point 67°C (153°F)
Refractive Index 1.39
Quantity 5 g
UN Number UN1760
Beilstein 1787023
Formula Weight 218.56
Percent Purity 94%
Chemical Name or Material Ethyl 2-chloro-4,4,4-trifluoroacetoacetate
Hazard Category H227-H314-H335
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.