Learn More
Ethyl 3-(dimethylamino)acrylate, 99%
Description
Ethyl 3-(dimethylamino)acrylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ethyl 3-(dimethylamino)acrylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
Solubility
Soluble in water 90 g/L (25°C).
Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 17°C to 18°C |
| Boiling Point | 118°C to 121°C (10 mmHg) |
| Refractive Index | 1.512 |
| Linear Formula | (CH3)2NCH=CHCO2CH2CH3 |
| Quantity | 50 g |
| Solubility Information | Soluble in water 90g/L (25°C). |
| Formula Weight | 143.19 |
| Percent Purity | 99% |
| Chemical Name or Material | Ethyl 3-(dimethylamino)acrylate |
Safety and Handling
| Hazard Category | H317 |
| Hazard Statement | GHS H Statement H317 May cause an allergic skin reaction. |
| Precautionary Statement | P261-P272-P280g-P302+P352-P333+P313-P363-P501c |
| EINECSNumber | 402-650-4 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.