Learn More
Ethyl 3-ethoxypropionate, 99+%
Description
Ethyl 3-ethoxypropionate is used in the synthesis of phenols and selective inhibitors of cyclin-dependant kinase 4/6 for novel cancer therapies. It is also used as a solvent to prepare polymers. Further, it is used in paints and coatings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ethyl 3-ethoxypropionate is used in the synthesis of phenols and selective inhibitors of cyclin-dependant kinase 4/6 for novel cancer therapies. It is also used as a solvent to prepare polymers. Further, it is used in paints and coatings.
Solubility
Miscible with most organic solvents. Slightly miscible with cold water.
Notes
Incompatible with strong oxidizing agents, metals and strong acids.

Specifications
Specifications
Melting Point | -100°C |
Density | 0.949 |
Boiling Point | 169°C to 171°C |
Flash Point | 59°C (142°F) |
Odor | Ester-like |
Refractive Index | 1.4065 |
Linear Formula | CH3CH2OCH2CH2CO2CH2CH3 |
Quantity | 1000 mL |
UN Number | UN3272 |
Solubility Information | Miscible with most organic solvents. Slightly miscible with cold water. |
Show More |
Safety and Handling
Hazard Category | H226 |
Hazard Statement | GHS H Statement H226 Flammable liquid and vapour. |
Precautionary Statement | P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S. |
EINECSNumber | 212-112-9 |
RTECSNumber | UF3325000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.