Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethyl 3-phenylglycidate, cis + trans, 90+%

Click to view available options
Quantity:
50mL
250mL
1000mL
121-39-1
C11H12O3
192.214
MFCD00005123
GOMAKLPNAAZVCJ-UHFFFAOYSA-N
ethyl 3-phenylglycidate,ethyl phenylglycidate,oxiranecarboxylic acid, 3-phenyl-, ethyl ester,ethyl 2,3-epoxy-3-phenylpropionate,ethyl 3-phenyloxiranecarboxylate,glycidic acid, 3-phenyl-, ethyl ester,2-ethyl-3-phenylglycidate,fema no. 2454,ccris 2623,ethyl3-phenylglycidate
8469
ethyl 3-phenyloxirane-2-carboxylate
CCOC(=O)C1C(O1)C2=CC=CC=C2
121-39-1
C11H12O3
192.214
MFCD00005123
GOMAKLPNAAZVCJ-UHFFFAOYSA-N
ethyl 3-phenylglycidate,ethyl phenylglycidate,oxiranecarboxylic acid, 3-phenyl-, ethyl ester,ethyl 2,3-epoxy-3-phenylpropionate,ethyl 3-phenyloxiranecarboxylate,glycidic acid, 3-phenyl-, ethyl ester,2-ethyl-3-phenylglycidate,fema no. 2454,ccris 2623,ethyl3-phenylglycidate
8469
ethyl 3-phenyloxirane-2-carboxylate
CCOC(=O)C1C(O1)C2=CC=CC=C2
Quantity 1000mL
Flash Point >110°C (230°F)
Boiling Point 95°C to 97°C (0.5mmHg)
Formula Weight 192.21
Percent Purity ≥90%
Chemical Name or Material Ethyl 3-phenylglycidate, cis + trans

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Recommended Storage : 2° to 8°C