Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 4-chloroacetoacetate, 98%, ACROS Organics™

Click to view available options
Quantity:
50g
250g
1kg
Packaging:
Glass bottle
638-07-3
C6H9ClO3
164.59
MFCD00000939
OHLRLMWUFVDREV-UHFFFAOYSA-N
ethyl 4-chloroacetoacetate,4-chloroacetoacetic acid ethyl ester,butanoic acid, 4-chloro-3-oxo-, ethyl ester,ethyl gamma-chloroacetoacetate,unii-5ae82250cm,ccris 6791,ethyl chloroacetyl acetate,butanoic acid, 4-chloro-3-oxo-ethylester,ethyl 4-chloro-3-oxobutyrate,ethyl 4-chloro-3-oxo-butanoate
69484
ethyl 4-chloro-3-oxobutanoate
CCOC(=O)CC(=O)CCl
638-07-3
C6H9ClO3
164.59
MFCD00000939
OHLRLMWUFVDREV-UHFFFAOYSA-N
ethyl 4-chloroacetoacetate,4-chloroacetoacetic acid ethyl ester,butanoic acid, 4-chloro-3-oxo-, ethyl ester,ethyl gamma-chloroacetoacetate,unii-5ae82250cm,ccris 6791,ethyl chloroacetyl acetate,butanoic acid, 4-chloro-3-oxo-ethylester,ethyl 4-chloro-3-oxobutyrate,ethyl 4-chloro-3-oxo-butanoate
69484
ethyl 4-chloro-3-oxobutanoate
CCOC(=O)CC(=O)CCl
Color Colorless to Red-Yellow
Flash Point 96°C
Melting Point -8.0°C
Packaging Glass bottle
Density 1.2100g/mL
Infrared Spectrum Authentic
Boiling Point 103.0°C (16.0 mmHg)
Refractive Index 1.4510 to 1.4530
Quantity 250g
Specific Gravity 1.21
Assay Percent Range 97.5% min. (GC)
Linear Formula ClCH2COCH2CO2C2H5
Beilstein 03,663
Solubility Information Solubility in water: 47.5g/L water (20°C). Other solubilities: soluble in ether,benzene,chloroform,,methylenechloride and carbontetrachloride
Formula Weight 164.59
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material Ethyl 4-chloroacetoacetate, 98%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Toxic if swallowed.
Toxic in contact with skin.
Toxic to aquatic life with long lasting effects.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 211-317-0

RTECSNumber : AK5110000

TSCA : TSCA