Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 4-Quinazolone-2-carboxylate 98.0+%, TCI America™
SDP

Catalog No. E11231G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E11231G 1 g
E11235G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E11231G Supplier TCI America Supplier No. E11231G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 29113-33-5
Molecular Formula C11H10N2O3
Molecular Weight (g/mol) 218.212
MDL Number MFCD00051583
InChI Key BMCAWNQKVVTNFP-UHFFFAOYSA-N
Synonym ethyl 4-quinazolone-2-carboxylate, ethyl 4-oxo-3,4-dihydroquinazoline-2-carboxylate, ethyl 4-hydroxyquinazoline-2-carboxylate, ethyl 4-oxo-3,4-dihydro-2-quinazolinecarboxylate, ethyl 4-oxo-3h-quinazoline-2-carboxylate, 4-quinazolone-2-carboxylic acid ethyl ester, 4-hydroxy-quinazoline-2-carboxylic acid ethyl ester, 2-quinazolinecarboxylic acid, 1,4-dihydro-4-oxo-, ethyl ester, acmc-20aic8, enamine_001988
PubChem CID 293295
IUPAC Name ethyl 4-oxo-1H-quinazoline-2-carboxylate
SMILES CCOC(=O)C1=NC(=O)C2=CC=CC=C2N1

Specifications

Melting Point 192°C
Color White
Quantity 1 g
Formula Weight 218.21
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 4-Quinazolone-2-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.