Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 5-methyl-4-oxo-3,4-dihydropyrrolo[2,1-f]-1,2,4-triazine-6-carboxylate, 97%, Thermo Scientific™

Catalog No. AAH6510103
Click to view available options
Quantity:
250 mg
1 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 427878-70-4
Molecular Formula C10H11N3O3
Molecular Weight (g/mol) 221.216
MDL Number MFCD11044884
InChI Key XCUHVGKOOKXZNK-UHFFFAOYSA-N
Synonym ethyl 5-methyl-4-oxo-1,4-dihydropyrrolo 2,1-f 1,2,4 triazine-6-carboxylate, ethyl 5-methyl-4-oxo-3,4-dihydropyrrolo 1,2-f 1,2,4 triazine-6-carboxylate, ethyl 4-hydroxy-5-methylpyrrolo 2,1-f 1,2,4 triazine-6-carboxylate, ethyl 4-hydroxy-5-methylpyrrolo 1,2-f 1,2,4 triazine-6-carboxylate, ethyl 5-methyl-4-oxo-3h,4h-pyrrolo 2,1-f 1,2,4 triazine-6-carboxylate, ethyl 5-methyl-4-oxo-3,4-dihydropyrrolo 2,1-f 1,2,4 triazine-6-carboxylate, ethyl 5-methyl-4-oxo-1h-pyrrolo 2,1-f 1,2,4 triazine-6-carboxylate, ethyl 5-methyl-4-oxo-3h-pyrrolo 2,1-f 1,2,4 triazine-6-carboxylate, 5-methyl-4-oxo-3,4-dihydro-pyrrolo 2,1-f 1,2,4 triazine-6-carboxylic acid ethyl ester, pyrrolo 2,1-f 1,2,4 triazine-6-carboxylic acid, 1,4-dihydro-5-methyl-4-oxo-, ethyl ester
PubChem CID 11572090
IUPAC Name ethyl 5-methyl-4-oxo-1H-pyrrolo[2,1-f][1,2,4]triazine-6-carboxylate
SMILES CCOC(=O)C1=CN2C(=C1C)C(=O)N=CN2

Specifications

Color Yellow
Quantity 1 g
Beilstein 10162668
Formula Weight 221.22
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Ethyl 5-methyl-4-oxo-3,4-dihydropyrrolo[2,1-f]-1,2,4-triazine-6-carboxylate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.