Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Alcohol, Denatured, Anhydrous, Reagents

Catalog No. 01337531
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-531 500 mL
01-337-533 20 L
01-337-532 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-337-531 Supplier Reagents Holdings Llc Supplier No. C2120600500A
Add to Cart
Add to Cart

Ethyl Alcohol, Denatured, Anhydrous, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 64-17-5 , 67-63-0 , 67-56-1 , 108-10-1
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym Denatured alcohol, ethyl alcohol
IUPAC Name ethanol
SMILES CCO

Specifications

Melting Point -90°C
Color Colorless
Quantity 500 mL
Packaging Poly Bottle
Physical Form Liquid
Specific Gravity 0.7893
Boiling Point 77°C
Grade Reagent
Chemical Name or Material Ethyl Alcohol, Denatured, Anhydrous
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.