Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl benzimidate hydrochloride, 97%

Catalog No. p-7047130
Click to view available options
Quantity:
10 g
50 g

CAS: 5333-86-8 | C9H12ClNO | 185.65 g/mol

Ethyl benzimidate hydrochloride is used in the preparation of (4R)-ethyl 2-phenyl-4,5-dihydrothiazole-4-carboxylate by reacting with (R)-ethyl cysteine hydrochloride. It also reacts with D-penicillamine methyl ester hydrochloride and triethylamine to prepare methyl-5,5- dimethyl-2-phenyl-2-thiazoline-4-carboxylate. Further, it is used as an intermediate for organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl benzimidate hydrochloride is used in the preparation of (4R)-ethyl 2-phenyl-4,5-dihydrothiazole-4-carboxylate by reacting with (R)-ethyl cysteine hydrochloride. It also reacts with D-penicillamine methyl ester hydrochloride and triethylamine to prepare methyl-5,5- dimethyl-2-phenyl-2-thiazoline-4-carboxylate. Further, it is used as an intermediate for organic synthesis.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5333-86-8
Molecular Formula C9H12ClNO
Molecular Weight (g/mol) 185.65
MDL Number MFCD00043246
InChI Key MODZVIMSNXSQIH-UHFFFAOYSA-N
Synonym ethyl benzimidate hydrochloride, benzenecarboximidic acid, ethyl ester, hydrochloride, ethyl benzenecarboximidate hydrochloride, benzimidic acid ethyl ester hcl, ethylbenzimidate hydrochloride, ethyl benzimidate hcl, ethyl benzylimidate hydrochloride, ethyl iminobenzoate hydrochloride, benzimido ethyl ether hydrochloride, benzimidoyl ethyl ether hydrochloride
PubChem CID 79249
IUPAC Name ethyl benzenecarboximidate;hydrochloride
SMILES [H+].[Cl-].CCOC(=N)C1=CC=CC=C1

Specifications

Melting Point 115°C to 117°C
Quantity 10 g
Beilstein 3697589
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 185.65
Percent Purity 97%
Chemical Name or Material Ethyl benzimidate hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 226-248-1
TSCA Yes
Recommended Storage Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.