Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl hydrogen succinate, 95%

Catalog No. p-7036757
Click to view available options
Quantity:
1 g
5 g
25 g

CAS: 1070-34-4 | C6H10O4 | 146.142 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1070-34-4
Molecular Formula C6H10O4
Molecular Weight (g/mol) 146.142
MDL Number MFCD00053346
InChI Key LOLKAJARZKDJTD-UHFFFAOYSA-N
Synonym monoethyl succinate, ethyl hydrogen succinate, mono-ethyl succinate, butanedioic acid, monoethyl ester, butanedioic acid, 1-ethyl ester, succinic acid monoethyl ester, monoethyl butanedioate, bernsteinsaureathylester, succinic acid monoethyl, succinic acid ethyl ester
PubChem CID 70610
IUPAC Name 4-ethoxy-4-oxobutanoic acid
SMILES CCOC(=O)CCC(=O)O

Specifications

Density 1.141
Boiling Point 248°C
Flash Point 113°C (235°F)
Refractive Index 1.432
Linear Formula HOOCCH2CH2CO2CH2CH3
Quantity 1 g
Formula Weight 146.14
Percent Purity 95%
Chemical Name or Material Ethyl hydrogen succinate
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 213-973-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.