Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Nonadecanoate 96.0+%, TCI America™
SDP

Catalog No. p-7133820 Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N04595G 5 g
N045925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N04595G Supplier TCI America Supplier No. N04595G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 18281-04-4
Molecular Formula C21H42O2
Molecular Weight (g/mol) 326.565
MDL Number MFCD00026690
InChI Key ICVYQLQYVCXJNE-UHFFFAOYSA-N
Synonym Nonadecanoic Acid Ethyl Ester
PubChem CID 29008
IUPAC Name ethyl nonadecanoate
SMILES CCCCCCCCCCCCCCCCCCC(=O)OCC

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 326.57
Percent Purity ≥96.0% (GC)
Physical Form Crystals
Chemical Name or Material Ethyl Nonadecanoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.