Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl propionate

Catalog No. AA15498D5
Change view
Click to view available options
Quantity:
1 kg
3 x 1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA15498D5 3 x 1 kg
AA15498A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA15498D5 Supplier Thermo Scientific Chemicals Supplier No. 015498D5
Only null left
Add to Cart

CAS: 105-37-3 | C5H10O2 | 102.133 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-37-3
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.133
MDL Number MFCD00009308
InChI Key FKRCODPIKNYEAC-UHFFFAOYSA-N
Synonym ethyl propionate, propanoic acid, ethyl ester, propionic ester, propionic ether, propionic acid, ethyl ester, ethylpropionate, propionate d'ethyle, propanoic acid ethyl ester, ethyl n-propionate, propionic acid ethyl ester
PubChem CID 7749
ChEBI CHEBI:41330
IUPAC Name ethyl propanoate
SMILES CCC(=O)OCC

Specifications

Melting Point -73°C
Density 0.891 g/mL
Boiling Point 99°C
Flash Point 12°C (54°F)
Refractive Index 1.385
Quantity 3 x 1 kg
UN Number UN1195
Beilstein 506287
Merck Index 14,3847
Formula Weight 102.13
Physical Form Liquid
Chemical Name or Material Ethyl propionate
Show More Show Less
Hazard Category H225
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHYL PROPIONATE
EINECSNumber 203-291-4
RTECSNumber UF3675000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.