Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl pyrrolidinoacetate, 98%, Thermo Scientific™

Catalog No. AC350920050
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle

Chemical Identifiers

CAS 22041-19-6
Molecular Formula C8H15NO2
Molecular Weight (g/mol) 157.213
MDL Number MFCD00012085
InChI Key IIIBKVMTKMLDFJ-UHFFFAOYSA-N
Synonym ethyl 2-pyrrolidin-1-yl acetate, ethyl 1-pyrrolidineacetate, ethylpyrrolidinoacetate, 1-pyrrolidineacetic acid, ethyl ester, ethyl pyrrolidinoacetate, ethyl pyrrolidin-1-ylacetate, ethyl n-pyrrolidinylacetate, ethyl 1-pyrrolidinylacetate, pyrrolidin-1-yl-acetic acid ethyl ester, ethyl-1-pyrrolidineacetate
PubChem CID 210996
IUPAC Name ethyl 2-pyrrolidin-1-ylacetate
SMILES CCOC(=O)CN1CCCC1

Specifications

Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Refractive Index 1.4460 to 1.4480 (20°C, 589nm)
Quantity 5 g
Beilstein 20, V,1, 335
Formula Weight 157.21
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl pyrrolidinoacetate, 98%
Show More Show Less
RTECSNumber UX9670300

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.