Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylcyclohexane, 99+%, Thermo Scientific Chemicals

Catalog No. p-128998
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1678-91-7
Molecular Formula C8H16
Molecular Weight (g/mol) 112.21
MDL Number MFCD00001523
InChI Key IIEWJVIFRVWJOD-UHFFFAOYSA-N
Synonym cyclohexane, ethyl, ethyl cyclohexane, cyclohexylethane, ethyl-cyclohexane, acmc-1c1qu, ethylcyclohexane, dsstox_cid_30334, dsstox_gsid_51779, wln: l6tj a2, qspl 100
PubChem CID 15504
IUPAC Name ethylcyclohexane
SMILES CCC1CCCCC1

Specifications

Melting Point -111°C
Color Colorless
Density 0.7800g/mL
Boiling Point 130°C to 132°C
Flash Point 18°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Refractive Index 1.4311 to 1.4331
Linear Formula C6H11C2H5
Quantity 25 mL
Beilstein 05, 35
Specific Gravity 0.78
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol,diethyl ether,acetone,,benzene,carbon tetrachloride,lignoin
Formula Weight 112.21
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Ethylcyclohexane, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
EINECSNumber 216-835-0
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.