Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ethyle4-chloro-2-(trifluoromethyl)pyrimidine-5-carboxylate, 97%, Thermo Scientific™

Catalog No. SPB04750DA
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SPB04750DA 1 g
SPB04750EA 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SPB04750DA Supplier Thermo Scientific Chemicals Supplier No. SPB04750DA
Add to Cart

Chemical Identifiers

CAS 720-01-4
Molecular Formula C8H6ClF3N2O2
Molecular Weight (g/mol) 254.59
MDL Number MFCD00173897
InChI Key DSULCDCBENGHAX-UHFFFAOYSA-N
Synonym ethyl 4-chloro-2-trifluoromethyl pyrimidine-5-carboxylate, 4-chloro-2-trifluoromethyl-pyrimidine-5-carboxylic acid ethyl ester, 5-pyrimidinecarboxylic acid, 4-chloro-2-trifluoromethyl-, ethyl ester, 4-chloro-2-trifluoromethyl-pyrimidine-5-carboxylicacidethylester, ethyl 2-trifluoromethyl-4-chloropyrimidine-5-carboxylate, ethyl 4-chloro-2-trifluoromethyl pyrimidine-5-car, ethyl 4-chloro-2-trifluoromethyl-pyrimidine-5-carboxylate, ethyl-4-chloro-2-trifluoromethyl pyrimidine-5-carboxylate, 5-pyrimidinecarboxylicacid, 4-chloro-2-trifluoromethyl-, ethyl ester
PubChem CID 2737166
IUPAC Name ethyl 4-chloro-2-(trifluoromethyl)pyrimidine-5-carboxylate
SMILES CCOC(=O)C1=CN=C(N=C1Cl)C(F)(F)F

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 254.59
Percent Purity 97%
Chemical Name or Material ethyle4-chloro-2-(trifluoromethyl)pyrimidine-5-carboxylate
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.