Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylenediamine, 99%

Catalog No. AC118420100
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass Bottle
Plastic drum

Catalog No. AC118420100

Supplier: Thermo Scientific Chemicals 118420100

Only null left
Add to Cart

CAS: 107-15-3 | C2H8N2 | 60.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-15-3
Molecular Formula C2H8N2
Molecular Weight (g/mol) 60.10
MDL Number MFCD00008204
InChI Key PIICEJLVQHRZGT-UHFFFAOYSA-N
Synonym ethylenediamine, 1,2-ethanediamine, 1,2-diaminoethane, ethylene diamine, ethylenediamine, dimethylenediamine, edamine, 1,2-ethylenediamine, aethaldiamin, aethylenediamin
PubChem CID 3301
ChEBI CHEBI:30347
IUPAC Name ethane-1,2-diamine
SMILES NCCN

Specifications

Melting Point 11.0°C
Color Colorless to Yellow
Density 0.8980g/mL
Boiling Point 117.0°C to 118.0°C
Flash Point 38°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic drum
Refractive Index 1.4555 to 1.4575
Linear Formula H2NCH2CH2NH2
Quantity 10 L
Beilstein 04, 230
Fieser 01,372; 04,231; 13,157
Specific Gravity 0.898
Merck Index 15, 3849
Solubility Information Solubility in water: freely soluble. Other solubilities: freely soluble in alcohol, slightly soluble in ether, soluble in benzene
Viscosity 1.6 mPa.s (20°C)
Formula Weight 60.1
Percent Purity 99%
Grade Extra Pure
Physical Form Liquid, Fuming In Air
Chemical Name or Material Ethylenediamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Toxic in contact with skin.
May cause an allergic skin reaction.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN (
EINECSNumber 203-468-6

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.