Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylene Glycol Mono-2-chloroethyl Ether 98.0+%, TCI America™
SDP

Catalog No. C0169100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0169100ML 100 mL
C016925ML 25 mL
C0169500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C0169100ML Supplier TCI America Supplier No. C0169100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 628-89-7
Molecular Formula C4H9ClO2
Molecular Weight (g/mol) 124.56
MDL Number MFCD00002870
InChI Key LECMBPWEOVZHKN-UHFFFAOYSA-N
Synonym 2-2-chloroethoxy ethanol, ethanol, 2-2-chloroethoxy, diglycol chlorohydrin, diglycol chlorhydrin, 2-chloroethyl 2-hydroxyethyl ether, monochloropolyoxyethylene, 2-2-chloroethoxy ethan-1-ol, unii-r2up818l4i, diethylene glycol monochlorohydrin, polyethylene glycol chloride 210
PubChem CID 12361
IUPAC Name 2-(2-chloroethoxy)ethan-1-ol
SMILES OCCOCCCl

Specifications

Color Yellow
Boiling Point 185°C
Quantity 100 mL
Formula Weight 124.56
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethylene Glycol Mono-2-chloroethyl Ether
EINECSNumber (2)-0447
RTECSNumber KK1350000
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.