Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylene Glycol Monoethyl Ether Acetate (Laboratory), Fisher Chemical

Catalog No. e1814 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
4 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
E181-4 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. E181-4
This item is not returnable. View return policy

Chemical Identifiers

CAS 111-15-9
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.16
MDL Number MFCD00009251
InChI Key SVONRAPFKPVNKG-UHFFFAOYSA-N
Synonym ethylene glycol monoethyl ether acetate, cellosolve acetate, oxitol acetate, ethoxyethyl acetate, oxytol acetate, ethylglycol acetate, ethyl cellosolve acetate, 1-acetoxy-2-ethoxyethane, ethylglykolacetat, octan etoksyetylu
PubChem CID 8095
IUPAC Name 2-ethoxyethyl acetate
SMILES CCOCCOC(C)=O

Specifications

Melting Point -61°C
Density .9750g/cm³
Boiling Point 156°C
Packaging Glass Bottle
Linear Formula CH3CH2OCH2CH2OOCCH3
Quantity 4 L
Vapor Pressure 2.34mmHg at 25°C
Formula Weight 132.16
Grade Laboratory
Physical Form Liquid
Chemical Name or Material Ethylene Glycol Monoethyl Ether Acetate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.