Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylenediamine Anhydrous (Certified), Fisher Chemical™

Catalog No. p-127666 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 mL
Packaging:
Glass Bottle

Catalog No. E479-500

Chemical Identifiers

CAS 107-15-3
Molecular Formula C2H8N2
Molecular Weight (g/mol) 60.10
MDL Number MFCD00008204
InChI Key PIICEJLVQHRZGT-UHFFFAOYSA-N
Synonym ethylenediamine, 1,2-ethanediamine, 1,2-diaminoethane, ethylene diamine, ethylenediamine, dimethylenediamine, edamine, 1,2-ethylenediamine, aethaldiamin, aethylenediamin
PubChem CID 3301
ChEBI CHEBI:30347
IUPAC Name ethane-1,2-diamine
SMILES NCCN

Specifications

Melting Point 8.5°C
Density .8980g/cm³
pH 11.9
Boiling Point 118°C
Flash Point 35°C
Assay Percent Range 98 to 101 %
Packaging Glass Bottle
Linear Formula NH2CH2CH2NH2
Quantity 500 mL
UN Number 1604
Vapor Pressure 12mmHg at 25°C
Formula Weight 60.1
Percent Purity 98 to 101%
Grade Certified
Physical Form Liquid
Chemical Name or Material Ethylenediamine Anhydrous
Show More Show Less
DOTInformation DOT Class 8, : Corrosive
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.