Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl propionate, 99+%

Catalog No. p-3757894
Click to view available options
Quantity:
25 mL
500 mL
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 105-37-3 | C5H10O2 | 102.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-37-3
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.13
MDL Number MFCD00009308
InChI Key FKRCODPIKNYEAC-UHFFFAOYSA-N
Synonym ethyl propionate, propanoic acid, ethyl ester, propionic ester, propionic ether, propionic acid, ethyl ester, ethylpropionate, propionate d'ethyle, propanoic acid ethyl ester, ethyl n-propionate, propionic acid ethyl ester
PubChem CID 7749
ChEBI CHEBI:41330
IUPAC Name ethyl propanoate
SMILES CCC(=O)OCC

Specifications

Acidity 1mg KOH/g max.
Melting Point -73°C
Color Colorless to Yellow
Density 0.8870g/mL
Boiling Point 99°C
Flash Point 12°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.3830 to 1.387
Linear Formula C2H5CO2C2H5
Quantity 25 mL
Beilstein 02, 240
Fieser 01,385
Specific Gravity 0.887
Merck Index 15, 3899
Solubility Information Solubility in water: 25g/L water (15°C). Other solubilities: miscible with alcohol,ether and propylene glycol
Viscosity <1 mPa.s (20°C)
Formula Weight 102.13
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Ethyl propionate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-291-4

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.