Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ETS1 associated protein II Antibody, Novus Biologicals™
SDP

Catalog No. NB018836 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.05 mg
0.1 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018836 0.025 mg
NBP21149105 0.05 mg
NBP211491 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB018836 Supplier Novus Biologicals Supplier No. NBP2114910.025MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ETS1 associated protein II Polyclonal specifically detects ETS1 associated protein II in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.

Specifications

Antigen ETS1 associated protein II
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA 1:500
Formulation Tris/Glycine buffer, pH 7.4-7.8, HEPES,BSA 0.5%, glycerol 30%. with 0.02% Sodium Azide
Gene Alias AD022, dJ30M3.3, EAP2, EAPII, EC 3.1.4.-, ETS1-associated protein 2, ETS1-associated protein II, MGC111021,5'-Tyr-DNA phosphodiesterase, MGC9099,5'-tyrosyl-DNA phosphodiesterase, TRAF and TNF receptor associated protein, TTRAPTRAF and TNF receptor-associated protein, Tyr-DNA phosphodiesterase 2, tyrosyl-DNA phosphodiesterase 2
Gene Symbols TDP2
Host Species Rabbit
Immunogen The synthetic peptide taken from human Tyrosyl-DNAphosphodiesterase (TDP2) protein corresponding to amino acids spanning 335-356. The peptide was conserved in several other species. The selected peptide was post-synthetically modified to achieve highest antigenicity before coupling to carrier protein using hetero bifunctional cross linker for immunogen preparation
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Signal Transduction, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 51567
Test Specificity The antibody labels a strong band of TDP2 of 45kDa in TDP12 samples and in several cell lines. The antibody also labels a diffuse band of around 95 and 115kDa, the identity of these bands is not known but may represent TDP2 splice variants.
Target Species Rat, Human, Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.