Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ETS1 associated protein II Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$203.51 - $715.20
Specifications
| Antigen | ETS1 associated protein II |
|---|---|
| Dilution | Western Blot 1:500, ELISA 1:500 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB018836
![]() |
Novus Biologicals
NBP2114910.025MG |
0.025 mg |
Each for $203.51
|
|
|||||
NBP21149105
![]() |
Novus Biologicals
NBP2114910.05MG |
0.05 mg |
Each for $394.26
|
|
|||||
NBP211491
![]() |
Novus Biologicals
NBP211491 |
0.1 mg |
Each for $715.20
|
|
|||||
Description
ETS1 associated protein II Polyclonal specifically detects ETS1 associated protein II in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.Specifications
| ETS1 associated protein II | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rat, Human, Mouse | |
| TDP2 | |
| IgG | |
| The antibody labels a strong band of TDP2 of 45kDa in TDP12 samples and in several cell lines. The antibody also labels a diffuse band of around 95 and 115kDa, the identity of these bands is not known but may represent TDP2 splice variants. |
| Western Blot 1:500, ELISA 1:500 | |
| Polyclonal | |
| Rabbit | |
| 51567 | |
| The synthetic peptide taken from human Tyrosyl-DNAphosphodiesterase (TDP2) protein corresponding to amino acids spanning 335-356. The peptide was conserved in several other species. The selected peptide was post-synthetically modified to achieve highest antigenicity before coupling to carrier protein using hetero bifunctional cross linker for immunogen preparation | |
| Primary |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title