Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ETS1 associated protein II Antibody, Novus Biologicals™
SDP

Catalog No. NBP146178 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP146178 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP146178 Supplier Novus Biologicals Supplier No. NBP146178
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ETS1 associated protein II Polyclonal specifically detects ETS1 associated protein II in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen ETS1 associated protein II
Applications Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunohistochemistry 1:100-1:500, Immunoprecipitation 2-5 ug/mg lysate, Immunohistochemistry-Paraffin 1:100-1:500
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. O95551
Gene Alias AD022, dJ30M3.3, EAP2, EAPII, EC 3.1.4.-, ETS1-associated protein 2, ETS1-associated protein II, MGC111021,5'-Tyr-DNA phosphodiesterase, MGC9099,5'-tyrosyl-DNA phosphodiesterase, TRAF and TNF receptor associated protein, TTRAPTRAF and TNF receptor-associated protein, Tyr-DNA phosphodiesterase 2, tyrosyl-DNA phosphodiesterase 2
Gene Symbols TDP2
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 50 and 100 of human Tyrosyl DNA Phosphodiesterase-2 using the numbering given in entry NP_057698.2 (GeneID 51567).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 51567
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.