Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ETS1 associated protein II Antibody (TDP2/1258), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB007635 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB007635 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB007635 Supplier Novus Biologicals Supplier No. NBP250071AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ETS1 associated protein II Monoclonal specifically detects ETS1 associated protein II in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence.

Specifications

Antigen ETS1 associated protein II
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Monoclonal
Clone TDP2/1258
Conjugate Alexa Fluor 405
Dilution Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias AD022, dJ30M3.3, EAP2, EAPII, EC 3.1.4.-, ETS1-associated protein 2, ETS1-associated protein II, MGC111021,5'-Tyr-DNA phosphodiesterase, MGC9099,5'-tyrosyl-DNA phosphodiesterase, TRAF and TNF receptor associated protein, TTRAPTRAF and TNF receptor-associated protein, Tyr-DNA phosphodiesterase 2, tyrosyl-DNA phosphodiesterase 2
Gene Symbols TDP2
Host Species Mouse
Immunogen Recombinant full-length human ETS1 associated protein II protein (Uniprot: O95551)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction, Transcription Factors and Regulators
Primary or Secondary Primary
Test Specificity This MAb recognizes a protein of 41kDa, which is identified as TDP2. It is a member of a superfamily of divalent cation-dependent phosphodiesterases. The encoded protein associates with CD40, tumor necrosis factor (TNF) receptor-75 and TNF receptor associated factors (TRAFs), and inhibits nuclear factor-kappa-B activation. This protein has sequence and structural similarities with APE1 endonuclease, which is involved in both DNA repair and the activation of transcription factors. DNA repair enzyme that can remove a variety of covalent adducts from DNA through hydrolysis of a 5'-phosphodiester bond, giving rise to DNA with a free 5' phosphate. Catalyzes the hydrolysis of dead-end complexes between DNA and the topoisomerase 2 (TOP2) active site tyrosine residue. Hydrolyzes 5'-phosphoglycolates on protruding 5' ends on DNA double-strand breaks (DSBs) due to DNA damage by radiation and free radicals. The 5'-tyrosyl DNA phosphodiesterase activity can enable the repair of TOP2-induced DSBs without the need for nuclease activity, creating a 'clean' DSB with 5'-phosphate termini that are ready for ligation. Has also 3'-tyrosyl DNA phosphodiesterase activity, but less efficiently and much slower than TDP1. May also act as a negative regulator of ETS1 and may inhibit nuclear factor-kappa-B activation.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.