Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ETS1 associated protein II Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP30333620UL
Description
ETS1 associated protein II Polyclonal specifically detects ETS1 associated protein II in Human, Mouse samples. It is validated for Western Blot, Knockout Validated.Specifications
ETS1 associated protein II | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, KnockDown | |
Unconjugated | |
AD022, dJ30M3.3, EAP2, EAPII, EC 3.1.4.-, ETS1-associated protein 2, ETS1-associated protein II, MGC111021,5'-Tyr-DNA phosphodiesterase, MGC9099,5'-tyrosyl-DNA phosphodiesterase, TRAF and TNF receptor associated protein, TTRAPTRAF and TNF receptor-associated protein, Tyr-DNA phosphodiesterase 2, tyrosyl-DNA phosphodiesterase 2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human ETS1 associated protein II (NP_057698.2). | |
20 μL | |
Signal Transduction, Transcription Factors and Regulators | |
51567 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction