Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ETS1 associated protein II Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$243.50 - $524.00

Specifications

Antigen ETS1 associated protein II
Applications Western Blot, KnockDown
Classification Polyclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB094725
SDP
View Documents
Novus Biologicals
NBP30333620UL
20 μL
Each for $243.50
Only null left
Add to Cart
 
NB094724
SDP
View Documents
Novus Biologicals
NBP303336100UL
100 μL
Each for $524.00
Only null left
Add to Cart
 
Description

Description

ETS1 associated protein II Polyclonal specifically detects ETS1 associated protein II in Human, Mouse samples. It is validated for Western Blot, Knockout Validated.
Specifications

Specifications

ETS1 associated protein II
Polyclonal
Purified
RUO
Human, Mouse
AD022, dJ30M3.3, EAP2, EAPII, EC 3.1.4.-, ETS1-associated protein 2, ETS1-associated protein II, MGC111021,5'-Tyr-DNA phosphodiesterase, MGC9099,5'-tyrosyl-DNA phosphodiesterase, TRAF and TNF receptor associated protein, TTRAPTRAF and TNF receptor-associated protein, Tyr-DNA phosphodiesterase 2, tyrosyl-DNA phosphodiesterase 2
Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human ETS1 associated protein II (NP_057698.2).
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Western Blot, KnockDown
Unconjugated
Rabbit
Signal Transduction, Transcription Factors and Regulators
PBS with 50% glycerol, pH7.3.
51567
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.