Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ETS2 Antibody (OTI2A3), DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP270694R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270694R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270694R Supplier Novus Biologicals Supplier No. NBP270694R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ETS2 Monoclonal specifically detects ETS2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen ETS2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI2A3
Conjugate DyLight 550
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias ETS2IT1, human erythroblastosis virus oncogene homolog 210protein C-ets-2, oncogene ETS-2, v-ets avian erythroblastosis virus E2 oncogene homolog 2, v-ets avian erythroblastosis virus E26 oncogene homolog 2, v-ets erythroblastosis virus E26 oncogene homolog 2 (avian)
Gene Symbols ETS2
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 90-349 of human ETS2 (NP_005230) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2114.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.