Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ETS2 Antibody (PCRP-ETS2-1D9), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB156741 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB156741 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB156741 Supplier Novus Biologicals Supplier No. NBP314209G
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

ETS2 Monoclonal antibody specifically detects ETS2 in Human samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence

Specifications

Antigen ETS2
Applications Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone PCRP-ETS2-1D9
Conjugate DyLight 488
Dilution Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias ETS2IT1, human erythroblastosis virus oncogene homolog 210protein C-ets-2, oncogene ETS-2, v-ets avian erythroblastosis virus E2 oncogene homolog 2, v-ets avian erythroblastosis virus E26 oncogene homolog 2, v-ets erythroblastosis virus E26 oncogene homolog 2 (avian)
Host Species Mouse
Immunogen Recombinant full-length human ETS2 protein (Uniprot: P15036)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2114
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.