Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ETV1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $648.50
Specifications
Antigen | ETV1 |
---|---|
Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
ETV1 Polyclonal specifically detects ETV1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
ETV1 | |
Polyclonal | |
Rabbit | |
Stem Cell Markers | |
DKFZp781L0674, ER81ets variant gene 1, ETS translocation variant 1, ets variant 1, Ets-related protein 81, MGC104699, MGC120533, MGC120534 | |
ETV1 | |
IgG | |
Affinity Purified |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human | |
2115 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:ASQSFPPPLMIKQEPRDFAYDSEVPSCHSIYMRQEGFLAHPSRTEGCMFEKGPRQFYDDTCVVPE | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title