Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ETV2/ER71 Antibody, Novus Biologicals™
SDP

Catalog No. NB10093393 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10093393 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10093393 Supplier Novus Biologicals Supplier No. NB10093393
Add to Cart
Add to Cart

Goat Polyclonal Antibody

ETV2/ER71 Polyclonal specifically detects ETV2/ER71 in Human samples. It is validated for Peptide ELISA.

Specifications

Antigen ETV2/ER71
Applications Peptide ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Peptide ELISA Detection limit 1:64000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. B9EIN1
Gene Alias ER71MGC129835, ETS translocation variant 2, ets variant 2, Ets-related protein 71, ETSRP71ets variant gene 2, MGC129834
Gene Symbols ETV2
Host Species Goat
Immunogen Peptide with sequence RCPKTNHRGPIQ corresponding to internal region according to NP_055024.2.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2116
Test Specificity Expected from sequence similarity: Human.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.