Learn More
Description
Specifications
Specifications
| Antigen | ETV3 |
| Applications | Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | DyLight 488 |
| Formulation | 50 mM Sodium Borate |
| Gene Alias | bA110J1.4, ETS domain transcriptional repressor PE1, ETS translocation variant 3, ets variant 3, ets variant gene 3, ETS family transcriptional repressor, METS, Mitogenic Ets transcriptional suppressor, PE-1ets variant gene 3, PE1FLJ79173 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human ETV3 fragment. |
| Purification Method | Antigen Affinity-purified |
| Quantity | 0.1 mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
