Learn More
Invitrogen™ ETV6 Polyclonal Antibody

Description
ETV6 Polyclonal Antibody for Western Blot, IHC (P), ELISA
Specifications
Specifications
| Antigen | ETV6 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 2.62 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 |
| Gene | Etv6 |
| Gene Accession No. | P41212, P97360 |
| Gene Alias | AW123102; AW557856; CGI-108; CSL4; Csl4p; ETS translocation variant 6; ETS variant 6; ets variant gene 6 (TEL oncogene); ETS-related protein Tel1; Etv6; hCsl4p; p13; RP11-452K12.9; SKI4; Ski4p; Tel; TEL/ABL; Tel1; TEL1 oncogene; THC5; Transcription factor ETV6 |
| Gene Symbols | Etv6 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.