Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ETV7 Antibody (OTI3B2), Novus Biologicals™
SDP

Catalog No. NBP245544 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP245544 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP245544 Supplier Novus Biologicals Supplier No. NBP245544
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ensure accurate, reproducible results in Western Blot

ETV7 Monoclonal specifically detects ETV7 in Human samples. It is validated for Western Blot.

Specifications

Antigen ETV7
Applications Western Blot
Classification Monoclonal
Clone OTI3B2
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000
Formulation PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide
Gene Alias ETS translocation variant 7, ets variant 7, ets variant gene 7 (TEL2 oncogene), TEL-2, TEL2 oncogene, TEL2ETS-related protein Tel2, TELBEts transcription factor TEL-2b, Tel-related Ets factor, transcription factor ets, transcription factor ETV7, Transcription factor Tel-2, TREF
Gene Symbols ETV7
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 90-341 of human ETV7 (NP_057219) produced in E.coli.
Molecular Weight of Antigen 38.8 kDa
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51513
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.