Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eukaryotic translation initiation factor 1 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB094702 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB094702 100 μL
NB094703 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB094702 Supplier Novus Biologicals Supplier No. NBP303239100UL

Rabbit Polyclonal Antibody

Eukaryotic translation initiation factor 1 Polyclonal specifically detects Eukaryotic translation initiation factor 1 in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen Eukaryotic translation initiation factor 1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias A121Protein translation factor SUI1 homolog, eIF1, EIF-1, EIF1A, eukaryotic translation initiation factor 1, ISO1, Sui1iso1, SUI1sui1iso1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-113 of human Eukaryotic translation initiation factor 1 (NP_005792.1).
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10209
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.