Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Euramco Hazardous Location Anti-Static Ducts

Safely conducts electricity to the ground circuit of the blower
Specifications
Color | Black |
---|---|
Diameter (English) Fan | 8 in. |
Diameter (Metric) Fan | 20.32 cm |
Product Type | Anti-static Duct |
Material | Conductive PVC |
Catalog Number | Mfr. No. | Length (English) | Length (English) Duct | Weight (English) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Length (English) | Length (English) Duct | Weight (English) | Price | Quantity & Availability | |||
19-817-741
![]() |
Euramco
FDT0815CBB |
15 ft. | 180 in. | 9 lb. | N/A | N/A | N/A | ||
19-817-743
![]() |
Euramco
FDT0825CBB |
25 ft. | 300 in. | 16 lb. | N/A | N/A | N/A | ||
Description
- Non-collapsible and reinforced
- Flame retardant
- Surface resistance less than 6 GΦ
Specifications
Black | |
20.32 cm | |
Conductive PVC |
8 in. | |
Anti-static Duct |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title