Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Euramco Ramfan™ EFi150XX Conductive Duct
SDP

For safe use in adverse atmospheres

Specifications

Color Black
Diameter (English) Fan 16 in.
Diameter (Metric) Fan 40.64 cm
Product Type Anti-static Duct
For Use With (Equipment) Ramfan electric blower
View More Specs

Products 2
Catalog Number Mfr. No. Length (English) Duct Length (Metric) Duct Price Quantity & Availability  
Catalog Number Mfr. No. Length (English) Duct Length (Metric) Duct Price Quantity & Availability  
19-822-707
SDP
View Documents
Euramco
FDT1615CBR
180 in. 457.2 cm N/A N/A N/A
19-817-744
SDP
View Documents
Euramco
FDT1625CBR
300 in. 762 cm N/A N/A N/A
Description

Description

  • High-strength
  • Lightweight anti-static glass reinforced ABS housing
  • IP55 rain tested switch enclosure
  • For hazardous or explosive gases safety
Specifications

Specifications

Black
40.64 cm
Ramfan electric blower
16 in.
Anti-static Duct
Conductive PVC
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.