Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Europium Mini Beads Antibody Conjugation Kit
Shop All R&D Systems Products

Click to view available options
Sufficient For:
10 Reactions
4 Reactions
Description
Kit Components:
- 4 or 10 Mini vials of 200nm Europium (dependent on kit)
- 1 vial of 1 x Reaction Buffer A
- 1 vial of 1 x Reaction Buffer B
- 1 vial of 10x Europium Quencher
- 1 vial of 1 x Resuspension Buffer
Specifications
Specifications
Content And Storage | -20°C |
Quantity | 40 μg |
Kit Contents | 4 or 10 Mini vials of 200nm Europium (dependent on kit), 1 vial of 1x Reaction Buffer A, 1 vial of 1x Reaction Buffer B, 1 vial of 10x Europium Quencher, 1 vial of 1x Resuspension Buffer |
Sufficient For | 10 Reactions |
For Use With (Application) | The optimum amount of antibody (which will influence the number of antibody molecules per particle) depends on the size of the particles (surface area) and on the application. You may need to conjugate different amounts of antibody to optimize your assay. We recommend testing with 0.1 mg/mL in the first instance, although slightly lower or higher concentrations can be explored to optimize performance in your particular application. Before testing antibody concentrations above 0.1 mg/mL we advise you to use our Antibody Concentration & Clean Up Kit for LATEX and EUROPIUM to remove interfering buffer components. This is due to the increase in contamination that will occur when conjugating using a larger volume of the stock antibody. Do not alter the reaction volume of 400 ul as this will reduce conjugation efficiency. |
Product Type | Europium Mini Antibody Labeling Kit |
For research use only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction