Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ EV SEC Columns
SDP

Catalog No. NB397878 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 Columns
5 Columns
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB397878 10 Columns
NB397877 5 Columns
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB397878 Supplier R&D Systems™ Supplier No. NBP31176310UNITS
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems.

Sample Type: Biofluids or cell culture supernatant

Performance

  • Separates extracellular vesicles (EVs) from circulating proteins
  • Maintains vesicle shape and functionality
  • Isolation time: approximately 15 to 20 minutes
  • Sample volume capacity: 500 uL to 2 mL

Storage: 4°C

Specifications

Quantity 10 Columns
Product Line Novus
Sample Type Biofluids or cell culture supernatant
Phase Size Exclusion
Technique SEC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.