Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EVA1/MPZL2 Antibody (OTI2C7), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP2713A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP2713A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP2713A30 Supplier Novus Biologicals Supplier No. NBP271553AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

EVA1/MPZL2 Monoclonal specifically detects EVA1/MPZL2 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen EVA1/MPZL2
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI2C7
Conjugate Alexa Fluor 350
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias EVAEpithelial V-like antigen 1EVA1myelin protein zero-like protein 2, myelin protein zero-like 2
Gene Symbols MPZL2
Host Species Mouse
Immunogen Full length recombinant protein of human EVA1/MPZL2 (NP_658911) produced in HEK293T cells.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 10205
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.