Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EVI-1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP152024 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP152024 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP152024 Supplier Novus Biologicals Supplier No. NBP152024
Add to Cart
Add to Cart

Goat Polyclonal Antibody

EVI-1 Polyclonal specifically detects EVI-1 in Human samples. It is validated for Western Blot, Peptide ELISA.

Specifications

Antigen EVI-1
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:16000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_001098547.3
Gene Alias AML1-EVI-1, AML1-EVI-1 fusion protein, ecotropic viral integration site 1, Ecotropic virus integration site 1 protein homolog, EVI-1, EVI1MDS1-EVI1, MDS1, MDS1 and EVI1 complex locus, MDS1 and EVI1 complex locus protein EVI1, MDS1 and EVI1 complex locus protein MDS1, MGC163392, MGC97004, myelodysplasia syndrome 1, Myelodysplasia syndrome 1 protein, Myelodysplasia syndrome-associated protein 1, oncogene EVI1, PRDM3, zinc finger protein Evi1
Gene Symbols MECOM
Host Species Goat
Immunogen Peptide with sequence C-DKESLHSTSH corresponding to internal region according to NP_001098547.3, NP_005232.2, NP_004982.2, NP_001157471.1, NP_001157472.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 2122
Test Specificity This antibody is expected to recognize all reported isoforms (NP_001098547.3; NP_005232.2; NP_004982.2; NP_001157471.1; NP_001157472.1). Reported variants represent identical protein: NP_001098548.2, NP_005232.2
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.