Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EVI2B Antibody (MEM-216), Biotin, Novus Biologicals™
SDP

Catalog No. NB002429 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002429 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB002429 Supplier Novus Biologicals Supplier No. NBP262207B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

EVI2B Monoclonal specifically detects EVI2B in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen EVI2B
Applications Flow Cytometry
Classification Monoclonal
Clone MEM-216
Conjugate Biotin
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias CD361 antigen, D17S376, ecotropic viral integration site 2B, Ecotropic viral integration site 2B protein homolog, EVDBprotein EVI2B, EVI-2B
Gene Symbols EVI2B
Host Species Mouse
Immunogen Raji cells
Purification Method Protein A purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 2124
Test Specificity The antibody BP53-12 recognizes defined epitope (aa 16-25) on human p53, a 50 kDa tumour suppressor found in increased amounts in a wide variety of transformed cells; it is frequently mutated or inactivated in many types of cancer.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.