Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EVI2B Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP159952

 View more versions of this product

Catalog No. NBP159952


Only null left
Add to Cart

Description

Description

EVI2B Polyclonal specifically detects EVI2B in Human samples. It is validated for Western Blot.
Specifications

Specifications

EVI2B
Polyclonal
Unconjugated
PBS, 2% Sucrose with 0.09% Sodium Azide
CD361 antigen, D17S376, ecotropic viral integration site 2B, Ecotropic viral integration site 2B protein homolog, EVDBprotein EVI2B, EVI-2B
Rabbit
Protein A purified
RUO
2124
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Western Blot
1 mg/ml
Western Blot 1.0 ug/ml
P34910
EVI2B
Synthetic peptides corresponding to EVI2B(ecotropic viral integration site 2B) The peptide sequence was selected from the middle region of EVI2B. Peptide sequence DICMDNIRENEISTKRTSIISLTPWKPSKSTLLADDLEIKLFESSENIED.
100 μL
Primary
Expected identity based on immunogen sequence: Human: 100%; Bovine: 92%; Mouse: 92%; Canine: 85%.
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.