Learn More
Thermo Scientific™ eVol™ XR Dispensing System Plungers
Catalog No. 66002041 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
eVol System, 1000 μL
eVol System, 50 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
66-002-041 eVol System, 1000 μL
66-002-030 eVol System, 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 66-002-041 Supplier Thermo Scientific™ Supplier No. 66002041
Order these plungers for use with the respective size syringe on the Thermo Scientific™ eVol­™ Dispensing System.
Use these Thermo Scientific™ eVol™ Dispensing System Plungers with the respective size syringe on the eVol Dispensing System. Like the syringes, these plungers are available in 5 to 1000μL sizes. The unique, hand-held eVol Dispensing System accurately and reproducibly performs a wide variety of liquid handling procedures. The easy-to-use device couples an analytical syringe to a programmable electronic controller, enabling highly reproducible dispensing for all users.
The eVol Dispensing System is ideal for laboratories that need to repeatedly dispense nonaqueous liquids, offering greater accuracy and reproducibility for applications such as:
  • Routine dispensing of volatile solvents, hazardous, corrosive or viscous chemicals
  • Preparation of calibration standards
  • Addition of internal standards
The system unifies two precision devices: a digitally controlled electronic drive, and a Thermo Scientific™ XCHANGE™ enabled analytical syringe.
Repeatable Performance for Higher Confidence in Your Results
  • Can be calibrated easily by a user to ensure validity of results
  • Intuitive, easy-to-use interface with touch wheel and full-color screen
  • Complies with GLP and GMP guidelines
Exceptional Versatility and Functionality
  • Integrated XCHANGE coupling allows syringes to be quickly and easily changed, preventing possible cross-contamination
  • Addition of Thermo Scientific™ MEPS™ (Micro Extraction by Packed Sorbent) technology to an XCHANGE syringe enables automated sample preparation by SPE in a hand-held system
TRUSTED_SUSTAINABILITY
Specifications
Product Type Dispensing System Plunger
Product Line HyperSep
For Use With (Equipment) eVol System, 1000 μL