Learn More
Invitrogen™ EVOS™ Light Cube, CFP
Click to view available options
Quantity:
1 each
Description
- Ideal for use with EVOS FL Color Imaging System, EVOS FL Auto Imaging System or EVOS FL Imaging Systems
- Each cube contains an LED, condensing optics, and hard-coated filters
- Continuous illumination enables consistent results 50,000+ hour bulb lifetime that affords lower operating costs
- Adjustable light intensity that helps reduce photobleaching User changeable and automatically recognized by the system
Specifications
Specifications
Type | CFP Fluorescent Light Cube |
Product Line | EVOS |
For Use With (Equipment) | Countess™ II FL Automated Cell Counter, EVOS™ M5000 Imaging System, EVOS™ M7000 Imaging System |
Includes | LED, Condensing Optics, Hard-coated Filters |
Wavelength | Ex: 445/45 Em: 510/42 |
Quantity | 1 each |