Learn More
EVOS™ EVOS™ Onstage Master Plate
Catalog No. 12563569
Click to view available options
Quantity:
1 Ea.
Allows user to utilize different types of vessels for imaging
  • Part of EVOS Onstage Incubator and holds EVOS Onstage Vessel Holder
  • Is an environmental chamber designed specifically for use with EVOS FL Auto Imaging System
  • Accommodates various types of cell culture vessels
  • Provides flexibility to extended time-lapse imaging experiments under precisely controlled environmental conditions
TRUSTED_SUSTAINABILITY
Specifications
Type Master Plate
Product Line EVOS
For Use With (Equipment) EVOS™ FL Auto Imaging System
Holds Onstage Vessel
Quantity 1 Ea.
WARNING: Cancer - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.