Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ Evotech™ Standard Full Body Harnesses

Catalog No. 17200118 Shop All MSA Products
Change view
Click to view available options
Buckle Style (Leg):
Quick-Connect
Tongue-buckle leg strap
Certifications/Compliance:
ANSI Z359.1, A10.32, CSA 259.10-06, OSHA
CSA Z259.10-06; Meets OSHA and ANSI Z359.1
D-Ring Location:
Back
Back, Side
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Buckle Style (Leg) Certifications/Compliance D-Ring Location
17-200-118 Quick-Connect CSA Z259.10-06; Meets OSHA and ANSI Z359.1 Back
17-200-116 Tongue-buckle leg strap ANSI Z359.1, A10.32, CSA 259.10-06, OSHA Back
17-200-117 Tongue-buckle leg strap ANSI Z359.1, A10.32, CSA 259.10-06, OSHA Back, Side
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17-200-118 Supplier MSA™ Supplier No. 10105940

Exceptional comfort, ease of use, durability and user safety

  • No binding edge on shoulder padding to prevent neck chafing
  • Breathable padding with moisture-wicking material keeps users cooler
  • Leg strap design keeps leg straps in place to increase mobility and comfort
  • Variable width sub-pelvic webbing provides 50% more surface area for better weight distribution
  • Dual-color webbing (red pattern outside, black inside) simplifies harness donning
  • Web management system conveniently stores excess webbing
  • Positionable lanyard clips
  • RFID enabled to allow easy tracking and inspection

Certifications

CSA Z259.10-06

Compliance

Meets OSHA and ANSI Z359.1

Specifications

Color Red
Buckle Style (Chest) Quick-Connect
Style Subpelvic full body
Webbing Material Polyester
For Use With (Application) Construction
Size Standard
Includes Manual
Certifications/Compliance CSA Z259.10-06; Meets OSHA and ANSI Z359.1
D-Ring Location Back
Buckle Style (Leg) Quick-Connect
Product Type Subpelvic Full-body Harness
Capacity (English) 400 lb.
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.